Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008396.2 | NP_001008397.1 | 15 - 16 | 2057 | 91 |
NM_007086.3 | NP_009017.1 | 16 - 17 | 2150 | 91 | |
XM_006720012.2 | XP_006720075.1 | 16 - 17 | 2148 | 91 | |
XM_011536373.2 | XP_011534675.1 | 15 - 16 | 1949 | 91 | |
XM_017020940.1 | XP_016876429.1 | 15 - 16 | 2061 | 91 | |
XM_017020941.1 | XP_016876430.1 | 13 - 14 | 1759 | 91 | |
GenBank mRNA | AJ006266.1 | - | 15 - 16 | 2099 | 91 |
AK001538.1 | - | 4 - 5 | 704 | 91 | |
AK001585.1 | - | 6 - 7 | 1073 | 91 | |
AK293005.1 | - | 16 - 17 | 2117 | 91 | |
BC063041.1 | - | 16 - 17 | 2126 | 91 |