Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008396.2 | NP_001008397.1 | 19 - 20 | 2646 | 105 |
NM_007086.3 | NP_009017.1 | 20 - 21 | 2739 | 105 | |
XM_011536373.2 | XP_011534675.1 | 19 - 20 | 2538 | 105 | |
GenBank mRNA | AJ006266.1 | - | 19 - 20 | 2688 | 105 |
AK001538.1 | - | 8 - 9 | 1293 | 105 | |
AK001585.1 | - | 10 - 11 | 1662 | 105 | |
AK001620.1 | - | 3 - 4 | 466 | 105 | |
AK293005.1 | - | 20 - 21 | 2706 | 105 | |
BC063041.1 | - | 20 - 21 | 2715 | 105 |