Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017917.3 | NP_060387.2 | 1 - 2 | 632 | 141 |
XM_005267782.3 | XP_005267839.1 | 2 - 3 | 513 | 141 | |
XM_017021388.1 | XP_016876877.1 | 2 - 3 | 513 | 141 | |
GenBank mRNA | AK000651.1 | - | 1 - 2 | 325 | 141 |
AK225447.1 | - | 1 - 2 | 325 | 141 | |
AK225512.1 | - | 1 - 2 | 325 | 141 | |
AK310700.1 | - | 1 - 2 | 80 | 141 | |
AY157304.1 | - | 1 - 2 | 403 | 141 | |
AY157305.1 | - | 1 - 2 | 403 | 141 | |
BC006823.1 | - | 1 - 2 | 83 | 141 | |
BC010293.2 | - | 1 - 2 | 70 | 141 | |
BC012563.1 | - | 1 - 2 | 64 | 141 | |
BX248043.1 | - | 2 - 3 | 443 | 141 |