Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000255.3 | NP_000246.2 | 7 - 8 | 1707 | 126 |
XM_005249143.3 | XP_005249200.1 | 7 - 8 | 1612 | 126 | |
GenBank mRNA | AB208845.1 | - | 2 - 3 | 285 | 126 |
AK292568.1 | - | 7 - 8 | 1683 | 126 | |
AK312611.1 | - | 7 - 8 | 1682 | 126 | |
BC016282.1 | - | 7 - 8 | 1570 | 126 | |
BT007434.1 | - | 6 - 7 | 1442 | 126 | |
BX647789.1 | - | 7 - 8 | 1532 | 126 | |
KF030882.1 | - | 6 - 7 | 1442 | 126 | |
KU178201.1 | - | 6 - 7 | 1442 | 126 | |
KU178202.1 | - | 6 - 7 | 1442 | 126 | |
M65131.1 | - | 7 - 8 | 1518 | 126 |