Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006965.3 | NP_008896.2 | 3 - 4 | 760 | 67 |
XM_005258341.3 | XP_005258398.1 | 3 - 4 | 747 | 67 | |
GenBank mRNA | AF038964.1 | - | 3 - 4 | 704 | 67 |
AK291246.1 | - | 3 - 4 | 739 | 67 | |
BC003566.1 | - | 3 - 4 | 710 | 67 | |
BC016801.1 | - | 3 - 4 | 731 | 67 | |
BT006658.1 | - | 2 - 3 | 569 | 67 | |
CR450328.1 | - | 2 - 3 | 569 | 67 | |
U68536.1 | - | 3 - 4 | 733 | 67 |