Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018349.3 | NP_060819.3 | 18 - 19 | 2315 | 114 |
XM_005254955.3 | XP_005255012.1 | 19 - 20 | 2796 | 114 | |
XM_005254960.2 | XP_005255017.1 | 11 - 12 | 1387 | 114 | |
XM_006720603.2 | XP_006720666.1 | 20 - 21 | 2552 | 114 | |
XM_011521770.1 | XP_011520072.1 | 17 - 18 | 2192 | 114 | |
XM_011521771.2 | XP_011520073.1 | 20 - 21 | 2721 | 114 | |
XM_011521772.2 | XP_011520074.1 | 19 - 20 | 2611 | 114 | |
XM_011521775.2 | XP_011520077.1 | 12 - 13 | 2067 | 114 | |
XM_017022403.1 | XP_016877892.1 | 17 - 18 | 2279 | 114 | |
GenBank mRNA | AK001953.1 | - | 2 - 3 | 134 | 114 |
AK002037.1 | - | 11 - 12 | 1090 | 114 | |
AY656717.1 | - | 18 - 19 | 2250 | 114 |