Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099402.1 | NP_001092872.1 | 6 - 7 | 705 | 69 |
XM_005268154.4 | XP_005268211.1 | 6 - 7 | 830 | 69 | |
XM_011537275.2 | XP_011535577.1 | 6 - 7 | 1002 | 69 | |
GenBank mRNA | AK311075.1 | - | 6 - 7 | 683 | 69 |
BC015935.1 | - | 6 - 7 | 684 | 69 | |
BQ432222.1 | - | 3 - 4 | 281 | 69 | |
BT006950.1 | - | 5 - 6 | 575 | 69 |