Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007123.5 | NP_009054.5 | 10 - 11 | 2231 | 87 |
NM_206933.2 | NP_996816.2 | 10 - 11 | 2231 | 87 | |
GenBank mRNA | AF055580.2 | - | 10 - 11 | 2231 | 87 |
AY481573.1 | - | 10 - 11 | 2231 | 87 |