Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007123.5 | NP_009054.5 | 5 - 6 | 1235 | 118 |
NM_206933.2 | NP_996816.2 | 5 - 6 | 1235 | 118 | |
GenBank mRNA | AF055580.2 | - | 5 - 6 | 1235 | 118 |
AY481573.1 | - | 5 - 6 | 1235 | 118 |