Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007123.5 | NP_009054.5 | 8 - 9 | 1942 | 64 |
NM_206933.2 | NP_996816.2 | 8 - 9 | 1942 | 64 | |
GenBank mRNA | AF055580.2 | - | 8 - 9 | 1942 | 64 |
AY481573.1 | - | 8 - 9 | 1942 | 64 |