Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001567.3 | NP_001558.3 | 16 - 17 | 2098 | 60 |
XM_005273978.3 | XP_005274035.1 | 17 - 18 | 2475 | 60 | |
XM_005273979.4 | XP_005274036.1 | 17 - 18 | 2322 | 60 | |
XM_011544999.2 | XP_011543301.1 | 17 - 18 | 2256 | 60 | |
XM_011545000.2 | XP_011543302.1 | 17 - 18 | 2322 | 60 | |
XM_017017673.1 | XP_016873162.1 | 18 - 19 | 2413 | 60 | |
GenBank mRNA | BC140853.1 | - | 16 - 17 | 2098 | 60 |
L36818.1 | - | 15 - 16 | 2075 | 60 | |
Y14385.1 | - | 16 - 17 | 2162 | 60 |