Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001567.3 | NP_001558.3 | 23 - 24 | 2802 | 99 |
XM_005273978.3 | XP_005274035.1 | 24 - 25 | 3179 | 99 | |
XM_005273979.4 | XP_005274036.1 | 24 - 25 | 3026 | 99 | |
XM_011544999.2 | XP_011543301.1 | 24 - 25 | 2960 | 99 | |
XM_011545000.2 | XP_011543302.1 | 24 - 25 | 3026 | 99 | |
XM_017017673.1 | XP_016873162.1 | 25 - 26 | 3117 | 99 | |
GenBank mRNA | BC140853.1 | - | 23 - 24 | 2802 | 99 |
L36818.1 | - | 22 - 23 | 2779 | 99 | |
Y14385.1 | - | 23 - 24 | 2866 | 99 |