Assay Design



Gene Transcripts

Gene Symbol
PPP2R5B
Entrez Gene ID
5526
Gene Name
protein phosphatase 2 regulatory subunit B'beta
Gene Aliases
B56B, PR61B
Chromosome Location
Chr.11: 64917553 - 64934478 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011545132.2 XP_011543434.1 1 - 2 80 64
XM_011545133.1 XP_011543435.1 1 - 2 82 64

View Full Product Details