Assay Design



Gene Transcripts

Gene Symbol
PGM1
Entrez Gene ID
5236
Gene Name
phosphoglucomutase 1
Gene Aliases
CDG1T, GSD14
Chromosome Location
Chr.1: 63593276 - 63660245 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172818.1 NP_001166289.1 5 - 6 1169 83
NM_001172819.1 NP_001166290.1 5 - 6 960 83
NM_002633.2 NP_002624.2 5 - 6 1083 83
GenBank mRNA AK293298.1 - 5 - 6 960 83
AK294186.1 - 6 - 7 1042 83
AK298505.1 - 5 - 6 890 83
AK312254.1 - 5 - 6 948 83
BC001756.2 - 5 - 6 996 83
BC019920.1 - 5 - 6 1083 83
BC067763.1 - 5 - 6 961 83
BC090856.1 - 5 - 6 928 83
BT006961.1 - 5 - 6 870 83
M83088.1 - 5 - 6 932 83

View Full Product Details