Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321831.1 | NP_001308760.1 | 8 - 9 | 879 | 71 |
NM_139239.2 | NP_640332.1 | 8 - 9 | 1042 | 71 | |
XM_005259340.3 | XP_005259397.1 | 8 - 9 | 1258 | 71 | |
XM_011527419.2 | XP_011525721.1 | 8 - 9 | 930 | 71 | |
XM_011527420.2 | XP_011525722.1 | 8 - 9 | 941 | 71 | |
XM_011527421.2 | XP_011525723.1 | 7 - 8 | 774 | 71 | |
XM_011527424.2 | XP_011525726.1 | 8 - 9 | 886 | 71 | |
XM_017027389.1 | XP_016882878.1 | 8 - 9 | 1261 | 71 | |
XM_017027390.1 | XP_016882879.1 | 8 - 9 | 1240 | 71 | |
XM_017027391.1 | XP_016882880.1 | 8 - 9 | 1193 | 71 | |
XM_017027392.1 | XP_016882881.1 | 8 - 9 | 838 | 71 | |
XM_017027393.1 | XP_016882882.1 | 8 - 9 | 837 | 71 | |
XM_017027394.1 | XP_016882883.1 | 6 - 7 | 1085 | 71 | |
XM_017027395.1 | XP_016882884.1 | 6 - 7 | 1067 | 71 | |
XM_017027396.1 | XP_016882885.1 | 8 - 9 | 1218 | 71 | |
XM_017027397.1 | XP_016882886.1 | 6 - 7 | 1024 | 71 | |
GenBank mRNA | AF385433.1 | - | 8 - 9 | 932 | 71 |
AF385434.1 | - | 8 - 9 | 1030 | 71 | |
BC006273.2 | - | 1 - 2 | 60 | 71 | |
BC119646.1 | - | 5 - 6 | 554 | 71 | |
BC119647.1 | - | 5 - 6 | 554 | 71 |