Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004163.4 | NP_004154.2 | 1 - 2 | 225 | 90 |
XM_006722518.1 | XP_006722581.1 | 1 - 2 | 503 | 90 | |
XM_011526131.1 | XP_011524433.1 | 1 - 2 | 503 | 90 | |
GenBank mRNA | AF329499.1 | - | 1 - 2 | 225 | 90 |
AK314115.1 | - | 1 - 2 | 503 | 90 | |
BC027474.1 | - | 1 - 2 | 93 | 90 |