Assay Design



Gene Transcripts

Gene Symbol
CCDC68
Entrez Gene ID
80323
Gene Name
coiled-coil domain containing 68
Gene Aliases
SE57-1
Chromosome Location
Chr.18: 54901509 - 54959539 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143829.1 NP_001137301.1 11 - 12 1115 126
NM_025214.2 NP_079490.1 11 - 12 1218 126
XM_006722552.3 XP_006722615.1 11 - 12 1167 126
XM_011526198.2 XP_011524500.1 11 - 12 2227 126
XM_011526199.2 XP_011524501.1 11 - 12 1126 126
XM_011526200.2 XP_011524502.1 11 - 12 1279 126
XM_011526201.2 XP_011524503.1 11 - 12 2186 126
XM_011526203.2 XP_011524505.1 10 - 11 1053 126
XM_017026011.1 XP_016881500.1 10 - 11 1219 126
GenBank mRNA AF273051.1 - 11 - 12 1116 126
AK058097.1 - 11 - 12 1115 126
BC029508.1 - 11 - 12 1172 126

View Full Product Details