Assay Design



Gene Transcripts

Gene Symbol
CCDC68
Entrez Gene ID
80323
Gene Name
coiled-coil domain containing 68
Gene Aliases
SE57-1
Chromosome Location
Chr.18: 54901509 - 54959539 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143829.1 NP_001137301.1 9 - 10 957 126
NM_025214.2 NP_079490.1 9 - 10 1060 126
XM_006722552.3 XP_006722615.1 9 - 10 1009 126
XM_011526198.2 XP_011524500.1 9 - 10 2069 126
XM_011526199.2 XP_011524501.1 9 - 10 968 126
XM_011526200.2 XP_011524502.1 9 - 10 1121 126
XM_011526201.2 XP_011524503.1 9 - 10 2028 126
XM_011526202.1 XP_011524504.1 9 - 10 1060 126
XM_011526203.2 XP_011524505.1 8 - 9 895 126
XM_017026011.1 XP_016881500.1 8 - 9 1061 126
GenBank mRNA AF273051.1 - 9 - 10 958 126
AK058097.1 - 9 - 10 957 126
AK313793.1 - 7 - 8 803 126
BC029508.1 - 9 - 10 1014 126

View Full Product Details