Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000672.3 | NP_000663.1 | 6 - 7 | 921 | 71 |
NM_001102470.1 | NP_001095940.1 | 6 - 7 | 921 | 71 | |
NR_132990.1 | 4 - 5 | 562 | 71 | ||
GenBank mRNA | AK092768.1 | - | 6 - 7 | 921 | 71 |
AK298290.1 | - | 2 - 3 | 490 | 71 | |
AK316326.1 | - | 4 - 5 | 490 | 71 |