Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021817.2 | NP_068589.1 | 1 - 2 | 241 | 68 |
XM_017002020.1 | XP_016857509.1 | 2 - 3 | 153 | 68 | |
GenBank mRNA | AB049054.1 | - | 1 - 2 | 241 | 68 |
AK090712.1 | - | 1 - 2 | 102 | 68 | |
BC029864.1 | - | 1 - 2 | 38 | 68 |