Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002410.4 | NP_002401.1 | 10 - 11 | 1519 | 99 |
XM_005263668.4 | XP_005263725.1 | 11 - 12 | 1884 | 99 | |
XM_005263669.4 | XP_005263726.1 | 11 - 12 | 1881 | 99 | |
XM_005263670.3 | XP_005263727.1 | 11 - 12 | 1537 | 99 | |
XM_006712534.3 | XP_006712597.1 | 12 - 13 | 1733 | 99 | |
XM_011511199.2 | XP_011509501.1 | 12 - 13 | 2168 | 99 | |
XM_011511200.1 | XP_011509502.1 | 11 - 12 | 1533 | 99 | |
XM_011511201.2 | XP_011509503.1 | 11 - 12 | 1637 | 99 | |
XM_011511202.1 | XP_011509504.1 | 11 - 12 | 1534 | 99 | |
XM_017004147.1 | XP_016859636.1 | 12 - 13 | 2008 | 99 | |
XM_017004148.1 | XP_016859637.1 | 11 - 12 | 1845 | 99 | |
GenBank mRNA | AF113921.1 | - | 10 - 11 | 1407 | 99 |
D17716.1 | - | 10 - 11 | 1519 | 99 |