Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002410.4 | NP_002401.1 | 4 - 5 | 717 | 115 |
XM_005263668.4 | XP_005263725.1 | 5 - 6 | 1082 | 115 | |
XM_005263669.4 | XP_005263726.1 | 5 - 6 | 1079 | 115 | |
XM_005263670.3 | XP_005263727.1 | 5 - 6 | 735 | 115 | |
XM_006712534.3 | XP_006712597.1 | 6 - 7 | 931 | 115 | |
XM_011511199.2 | XP_011509501.1 | 6 - 7 | 1366 | 115 | |
XM_011511200.1 | XP_011509502.1 | 5 - 6 | 731 | 115 | |
XM_011511201.2 | XP_011509503.1 | 5 - 6 | 835 | 115 | |
XM_011511202.1 | XP_011509504.1 | 5 - 6 | 732 | 115 | |
XM_011511204.2 | XP_011509506.1 | 6 - 7 | 934 | 115 | |
XM_017004147.1 | XP_016859636.1 | 6 - 7 | 1206 | 115 | |
XM_017004148.1 | XP_016859637.1 | 5 - 6 | 1043 | 115 | |
XM_017004149.1 | XP_016859638.1 | 5 - 6 | 1084 | 115 | |
GenBank mRNA | AF113921.1 | - | 4 - 5 | 605 | 115 |
D17716.1 | - | 4 - 5 | 717 | 115 |