Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012414.3 | NP_036546.2 | 26 - 27 | 3255 | 111 |
GenBank mRNA | AB020646.2 | - | 26 - 27 | 3091 | 111 |
AF004828.1 | - | 26 - 27 | 3133 | 111 | |
AF255648.1 | - | 26 - 27 | 3164 | 111 | |
AK027485.1 | - | 8 - 9 | 1177 | 111 | |
AK291234.1 | - | 26 - 27 | 3206 | 111 | |
AL117631.1 | - | 3 - 4 | 304 | 111 | |
BC146760.1 | - | 26 - 27 | 3091 | 111 |