Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012414.3 | NP_036546.2 | 30 - 31 | 3495 | 66 |
GenBank mRNA | AB020646.2 | - | 30 - 31 | 3331 | 66 |
AF004828.1 | - | 30 - 31 | 3373 | 66 | |
AF255648.1 | - | 30 - 31 | 3404 | 66 | |
AK291234.1 | - | 30 - 31 | 3446 | 66 | |
AL117631.1 | - | 7 - 8 | 544 | 66 | |
BC131573.1 | - | 30 - 31 | 3374 | 66 | |
BC146760.1 | - | 30 - 31 | 3331 | 66 |