Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021953.3 | NP_068772.2 | 2 - 3 | 784 | 121 |
NM_202002.2 | NP_973731.1 | 2 - 3 | 784 | 121 | |
NM_202003.2 | NP_973732.1 | 2 - 3 | 784 | 121 | |
XM_005253676.3 | XP_005253733.1 | 2 - 3 | 784 | 121 | |
XM_011520931.2 | XP_011519233.1 | 2 - 3 | 784 | 121 | |
XM_011520932.1 | XP_011519234.1 | 2 - 3 | 784 | 121 | |
XM_011520933.1 | XP_011519235.1 | 2 - 3 | 784 | 121 | |
XM_011520934.2 | XP_011519236.1 | 2 - 3 | 784 | 121 | |
XM_011520935.1 | XP_011519237.1 | 2 - 3 | 784 | 121 | |
GenBank mRNA | AY542306.1 | - | 1 - 2 | 552 | 121 |
BC006192.2 | - | 2 - 3 | 669 | 121 | |
BC006529.2 | - | 2 - 3 | 669 | 121 | |
BC012863.1 | - | 1 - 2 | 520 | 121 | |
BT006986.1 | - | 1 - 2 | 501 | 121 | |
U74612.1 | - | 2 - 3 | 615 | 121 | |
U74613.1 | - | 2 - 3 | 608 | 121 | |
U83113.1 | - | 2 - 3 | 570 | 121 |