Assay Design



Gene Transcripts

Gene Symbol
ERAP2
Entrez Gene ID
64167
Gene Name
endoplasmic reticulum aminopeptidase 2
Gene Aliases
L-RAP, LRAP
Chromosome Location
Chr.5: 96875940 - 96919703 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011543544.1 XP_011541846.1 5 - 6 1155 103
GenBank mRNA AB109031.1 - 5 - 6 1122 103
AB163917.1 - 5 - 6 1122 103
AF191545.1 - 4 - 5 1094 103
AK074281.1 - 5 - 6 1199 103
AK312864.1 - 5 - 6 1135 103
AY028805.1 - 4 - 5 1052 103
BC065240.1 - 4 - 5 981 103

View Full Product Details