Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018424.3 | NP_060894.2 | 8 - 9 | 1357 | 81 |
NM_019114.4 | NP_061987.3 | 8 - 9 | 1357 | 81 | |
XM_011518790.1 | XP_011517092.1 | 8 - 9 | 854 | 81 | |
XM_011518791.2 | XP_011517093.1 | 8 - 9 | 839 | 81 | |
XM_011518792.2 | XP_011517094.1 | 8 - 9 | 839 | 81 | |
XM_011518794.2 | XP_011517096.1 | 8 - 9 | 839 | 81 | |
XM_017014813.1 | XP_016870302.1 | 8 - 9 | 839 | 81 | |
XM_017014814.1 | XP_016870303.1 | 8 - 9 | 839 | 81 | |
XM_017014815.1 | XP_016870304.1 | 8 - 9 | 839 | 81 | |
GenBank mRNA | AB032179.3 | - | 8 - 9 | 1357 | 81 |
AF153416.1 | - | 8 - 9 | 1129 | 81 | |
AF153418.1 | - | 8 - 9 | 1129 | 81 | |
AK000388.1 | - | 8 - 9 | 741 | 81 |