Assay Design



Gene Transcripts

Gene Symbol
CGN
Entrez Gene ID
57530
Gene Name
cingulin
Gene Aliases
-
Chromosome Location
Chr.1: 151510859 - 151538692 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020770.2 NP_065821.1 16 - 17 3140 70
XM_005245365.4 XP_005245422.1 16 - 17 3152 70
GenBank mRNA AB037740.1 - 16 - 17 3113 70
AF263462.1 - 16 - 17 3147 70
AK290007.1 - 16 - 17 3113 70
BC146657.1 - 15 - 16 3418 70
BC150604.1 - 15 - 16 3019 70
BC152445.1 - 16 - 17 3113 70
BX647343.1 - 1 - 2 994 70

View Full Product Details