Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282196.1 | NP_001269125.1 | 2 - 3 | 329 | 118 |
NM_001282197.1 | NP_001269126.1 | 2 - 3 | 329 | 118 | |
XM_017029939.1 | XP_016885428.1 | 2 - 3 | 304 | 118 | |
GenBank mRNA | AK309433.1 | - | 2 - 3 | 263 | 118 |