Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321389.1 | NP_001308318.1 | 3 - 4 | 603 | 77 |
NM_001321390.1 | NP_001308319.1 | 3 - 4 | 248 | 77 | |
NM_001321391.1 | NP_001308320.1 | 3 - 4 | 282 | 77 | |
NM_015680.5 | NP_056495.3 | 2 - 3 | 340 | 77 | |
NR_135628.1 | 3 - 4 | 282 | 77 | ||
NR_135629.1 | 2 - 3 | 340 | 77 | ||
GenBank mRNA | AF151815.1 | - | 3 - 4 | 507 | 77 |
AK075274.1 | - | 2 - 3 | 296 | 77 | |
AK314975.1 | - | 2 - 3 | 283 | 77 | |
AY007160.1 | - | 2 - 3 | 313 | 77 | |
BC001393.2 | - | 2 - 3 | 263 | 77 | |
BC012821.2 | - | 2 - 3 | 312 | 77 | |
BI912613.1 | - | 3 - 4 | 249 | 77 | |
CR457401.1 | - | 2 - 3 | 175 | 77 | |
CX166359.1 | - | 3 - 4 | 599 | 77 | |
DB477110.1 | - | 2 - 3 | 327 | 77 | |
HY106884.1 | - | 2 - 3 | 340 | 77 |