Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039547.2 | NP_001034636.1 | 9 - 10 | 970 | 99 |
XM_011512624.2 | XP_011510926.1 | 9 - 10 | 768 | 99 | |
XM_017006088.1 | XP_016861577.1 | 9 - 10 | 955 | 99 | |
XM_017006089.1 | XP_016861578.1 | 10 - 11 | 1033 | 99 | |
GenBank mRNA | AK127641.1 | - | 9 - 10 | 938 | 99 |
BX648359.1 | - | 9 - 10 | 950 | 99 |