Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301206.1 | NP_001288135.1 | 2 - 3 | 427 | 80 |
NM_001717.3 | NP_001708.3 | 2 - 3 | 291 | 80 | |
XM_011521893.1 | XP_011520195.1 | 2 - 3 | 193 | 80 | |
GenBank mRNA | AK302992.1 | - | 3 - 4 | 307 | 80 |
HY045483.1 | - | 2 - 3 | 428 | 80 | |
L03427.1 | - | - | 287 | 80 |