Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301206.1 | NP_001288135.1 | 3 - 4 | 658 | 76 |
NM_001717.3 | NP_001708.3 | 3 - 4 | 522 | 76 | |
XM_011521893.1 | XP_011520195.1 | 3 - 4 | 424 | 76 | |
GenBank mRNA | AK302992.1 | - | 4 - 5 | 538 | 76 |
L03427.1 | - | - | 518 | 76 |