Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_028045.1 | - | 160 | ||
GenBank mRNA | AF047445.1 | - | - | 160 | |
AK290077.1 | - | - | 160 | ||
AK304677.1 | - | - | 160 | ||
AK310151.1 | - | - | 160 | ||
BC069672.1 | - | - | 160 | ||
BC069697.1 | - | - | 159 | ||
BC069734.1 | - | - | 160 |