Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000289.5 | NP_000280.1 | 18 - 19 | 2005 | 78 |
NM_001166686.1 | NP_001160158.1 | 20 - 21 | 2307 | 78 | |
NM_001166687.1 | NP_001160159.1 | 18 - 19 | 1859 | 78 | |
NM_001166688.1 | NP_001160160.1 | 17 - 18 | 1948 | 78 | |
XM_005268974.1 | XP_005269031.1 | 21 - 22 | 2204 | 78 | |
XM_005268975.1 | XP_005269032.1 | 21 - 22 | 2343 | 78 | |
XM_005268976.3 | XP_005269033.1 | 21 - 22 | 2386 | 78 | |
XM_005268977.1 | XP_005269034.1 | 20 - 21 | 2190 | 78 | |
XM_005268978.2 | XP_005269035.1 | 20 - 21 | 2103 | 78 | |
XM_005268979.1 | XP_005269036.1 | 20 - 21 | 2190 | 78 | |
XM_011538487.1 | XP_011536789.1 | 19 - 20 | 2045 | 78 | |
XM_011538488.2 | XP_011536790.1 | 18 - 19 | 1970 | 78 | |
XM_017019467.1 | XP_016874956.1 | 21 - 22 | 2273 | 78 | |
XM_017019468.1 | XP_016874957.1 | 20 - 21 | 2125 | 78 | |
XM_017019469.1 | XP_016874958.1 | 19 - 20 | 2097 | 78 | |
GenBank mRNA | AK126020.1 | - | 22 - 23 | 2660 | 78 |
AK126229.1 | - | 21 - 22 | 2257 | 78 | |
AK303681.1 | - | 13 - 14 | 1352 | 78 | |
AK308185.1 | - | 13 - 14 | 1328 | 78 | |
AK312284.1 | - | 18 - 19 | 1887 | 78 | |
BC000534.2 | - | 17 - 18 | 1835 | 78 | |
BC007798.2 | - | 16 - 17 | 1722 | 78 | |
BC012799.2 | - | 17 - 18 | 1835 | 78 | |
BC013298.2 | - | 17 - 18 | 1835 | 78 | |
BC021203.2 | - | 18 - 19 | 1857 | 78 | |
CR541698.1 | - | 8 - 9 | 875 | 78 | |
J05533.1 | - | 9 - 10 | 916 | 78 | |
KU178251.1 | - | 17 - 18 | 1820 | 78 | |
KU178252.1 | - | 16 - 17 | 1727 | 78 | |
KU178254.1 | - | 6 - 7 | 665 | 78 | |
M26066.1 | - | 18 - 19 | 1896 | 78 | |
U24183.1 | - | 17 - 18 | 2148 | 78 | |
Y00698.1 | - | 17 - 18 | 1839 | 78 |