Assay Design
Gene Transcripts
- Gene Symbol
- MEGF6
- Entrez Gene ID
-
1953
- Gene Name
- multiple EGF like domains 6
- Gene Aliases
-
EGFL3
- Chromosome Location
-
Chr.1: 3487942 - 3624757 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AB011539.2 |
- |
27 - 28 |
3654 |
153 |
View Full Product Details