Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271938.1 | NP_001258867.1 | 19 - 20 | 3985 | 62 |
NM_001410.2 | NP_001401.2 | 18 - 19 | 3784 | 62 | |
GenBank mRNA | AB011541.2 | - | 18 - 19 | 3784 | 62 |
AY280362.1 | - | 13 - 14 | 2107 | 62 | |
BC153880.1 | - | 18 - 19 | 3784 | 62 |