Assay Design



Gene Transcripts

Gene Symbol
PLCB4
Entrez Gene ID
5332
Gene Name
phospholipase C beta 4
Gene Aliases
ARCND2, PI-PLC
Chromosome Location
Chr.20: 9068710 - 9480816 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000933.3 NP_000924.3 17 - 18 1627 64
NM_182797.2 NP_877949.2 18 - 19 1690 64
XM_017027880.1 XP_016883369.1 19 - 20 1884 64
XM_017027881.1 XP_016883370.1 20 - 21 1866 64
XM_017027883.1 XP_016883372.1 17 - 18 1725 64
XM_017027886.1 XP_016883375.1 10 - 11 1052 64
GenBank mRNA AK025027.1 - 11 - 12 1108 64
AK307482.1 - 19 - 20 1752 64
BC117458.1 - 18 - 19 1645 64
BC143868.1 - 18 - 19 1645 64
BX648680.1 - 14 - 15 1327 64
L41349.1 - 11 - 12 1383 64

View Full Product Details