Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000933.3 | NP_000924.3 | 24 - 25 | 2502 | 136 |
NM_001172646.1 | NP_001166117.1 | 27 - 28 | 2754 | 136 | |
NM_182797.2 | NP_877949.2 | 25 - 26 | 2565 | 136 | |
XM_005260724.2 | XP_005260781.1 | 27 - 28 | 2795 | 136 | |
XM_005260728.2 | XP_005260785.1 | 18 - 19 | 1962 | 136 | |
XM_006723568.3 | XP_006723631.1 | 28 - 29 | 2777 | 136 | |
XM_006723569.2 | XP_006723632.1 | 27 - 28 | 2875 | 136 | |
XM_011529254.2 | XP_011527556.1 | 20 - 21 | 5897 | 136 | |
XM_017027880.1 | XP_016883369.1 | 26 - 27 | 2759 | 136 | |
XM_017027881.1 | XP_016883370.1 | 27 - 28 | 2741 | 136 | |
XM_017027882.1 | XP_016883371.1 | 23 - 24 | 2379 | 136 | |
XM_017027883.1 | XP_016883372.1 | 24 - 25 | 2600 | 136 | |
XM_017027884.1 | XP_016883373.1 | 25 - 26 | 2829 | 136 | |
XM_017027885.1 | XP_016883374.1 | 25 - 26 | 2506 | 136 | |
XM_017027886.1 | XP_016883375.1 | 17 - 18 | 1927 | 136 | |
GenBank mRNA | AK025027.1 | - | 18 - 19 | 1983 | 136 |
AK122699.1 | - | 27 - 28 | 2718 | 136 | |
AK307482.1 | - | 26 - 27 | 2627 | 136 | |
AK307837.1 | - | 27 - 28 | 2755 | 136 | |
BC117458.1 | - | 25 - 26 | 2520 | 136 | |
BC143868.1 | - | 25 - 26 | 2520 | 136 | |
BX648680.1 | - | 21 - 22 | 2202 | 136 | |
L41349.1 | - | 18 - 19 | 2258 | 136 |