Assay Design



Gene Transcripts

Gene Symbol
RFWD2
Entrez Gene ID
64326
Gene Name
ring finger and WD repeat domain 2
Gene Aliases
COP1, RNF200
Chromosome Location
Chr.1: 175944826 - 176207279 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001740.3 NP_001001740.1 9 - 10 1342 104
NM_001286644.1 NP_001273573.1 8 - 9 1217 104
NM_022457.6 NP_071902.2 10 - 11 1414 104
XM_005245447.2 XP_005245504.1 10 - 11 1437 104
XM_005245448.2 XP_005245505.1 9 - 10 1389 104
XM_006711487.2 XP_006711550.1 9 - 10 1380 104
XM_011509881.1 XP_011508183.1 11 - 12 1503 104
XM_017002059.1 XP_016857548.1 11 - 12 1503 104
XM_017002060.1 XP_016857549.1 11 - 12 1491 104
XM_017002061.1 XP_016857550.1 10 - 11 1449 104
XM_017002062.1 XP_016857551.1 10 - 11 1443 104
XM_017002063.1 XP_016857552.1 10 - 11 1437 104
XM_017002064.1 XP_016857553.1 10 - 11 1434 104
XM_017002065.1 XP_016857554.1 9 - 10 1389 104
XM_017002066.1 XP_016857555.1 9 - 10 1380 104
XM_017002067.1 XP_016857556.1 9 - 10 1377 104
XM_017002068.1 XP_016857557.1 9 - 10 1374 104
XM_017002069.1 XP_016857558.1 5 - 6 1017 104
XM_017002070.1 XP_016857559.1 10 - 11 1209 104
XM_017002071.1 XP_016857560.1 8 - 9 598 104
XM_017002072.1 XP_016857561.1 8 - 9 596 104
XM_017002073.1 XP_016857562.1 7 - 8 538 104
XM_017002074.1 XP_016857563.1 7 - 8 536 104
XM_017002075.1 XP_016857564.1 7 - 8 618 104
XM_017002076.1 XP_016857565.1 8 - 9 601 104
XM_017002077.1 XP_016857566.1 6 - 7 393 104
XM_017002078.1 XP_016857567.1 8 - 9 666 104
XM_017002079.1 XP_016857568.1 8 - 9 1081 104
XM_017002080.1 XP_016857569.1 4 - 5 320 104
GenBank mRNA AF508940.1 - 10 - 11 1148 104
AF527539.1 - 10 - 11 1411 104
AK001278.1 - 8 - 9 653 104
AK314750.1 - 10 - 11 1346 104
AY509921.1 - 9 - 10 1076 104
BC039723.1 - 8 - 9 664 104
BC094728.1 - 10 - 11 1402 104
KJ534928.1 - 10 - 11 1148 104
KJ534929.1 - 7 - 8 953 104
KJ534930.1 - 9 - 10 1088 104
KJ534931.1 - 9 - 10 1079 104
KJ534932.1 - 10 - 11 1136 104
KJ534933.1 - 4 - 5 647 104
KJ535061.1 - 7 - 8 853 104
KJ535076.1 - 9 - 10 1050 104

View Full Product Details