Assay Design



Gene Transcripts

Gene Symbol
RFWD2
Entrez Gene ID
64326
Gene Name
ring finger and WD repeat domain 2
Gene Aliases
COP1, RNF200
Chromosome Location
Chr.1: 175944826 - 176207279 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001740.3 NP_001001740.1 1 - 2 678 86
NM_001286644.1 NP_001273573.1 1 - 2 678 86
NM_022457.6 NP_071902.2 1 - 2 678 86
XM_005245447.2 XP_005245504.1 1 - 2 713 86
XM_005245448.2 XP_005245505.1 1 - 2 713 86
XM_006711487.2 XP_006711550.1 1 - 2 713 86
XM_011509881.1 XP_011508183.1 1 - 2 713 86
XM_017002059.1 XP_016857548.1 1 - 2 713 86
XM_017002060.1 XP_016857549.1 1 - 2 713 86
XM_017002061.1 XP_016857550.1 1 - 2 713 86
XM_017002062.1 XP_016857551.1 1 - 2 713 86
XM_017002063.1 XP_016857552.1 1 - 2 713 86
XM_017002064.1 XP_016857553.1 1 - 2 713 86
XM_017002065.1 XP_016857554.1 1 - 2 713 86
XM_017002066.1 XP_016857555.1 1 - 2 713 86
XM_017002067.1 XP_016857556.1 1 - 2 713 86
XM_017002068.1 XP_016857557.1 1 - 2 713 86
XM_017002069.1 XP_016857558.1 1 - 2 713 86
XM_017002070.1 XP_016857559.1 1 - 2 517 86
XM_017002072.1 XP_016857561.1 1 - 2 101 86
XM_017002074.1 XP_016857563.1 1 - 2 101 86
XM_017002075.1 XP_016857564.1 1 - 2 139 86
XM_017002078.1 XP_016857567.1 1 - 2 67 86
XM_017002079.1 XP_016857568.1 1 - 2 494 86
GenBank mRNA AF508940.1 - 1 - 2 412 86
AF527539.1 - 1 - 2 675 86
AK314750.1 - 1 - 2 610 86
AY509921.1 - 1 - 2 412 86
BC020845.1 - 1 - 2 585 86
BC039723.1 - 1 - 2 125 86
BC094728.1 - 1 - 2 666 86
KJ534928.1 - 1 - 2 412 86
KJ534929.1 - 1 - 2 412 86
KJ534930.1 - 1 - 2 412 86
KJ534931.1 - 1 - 2 412 86
KJ534932.1 - 1 - 2 412 86
KJ534933.1 - 1 - 2 412 86
KJ534992.1 - 1 - 2 412 86
KJ535061.1 - 1 - 2 412 86
KJ535076.1 - 1 - 2 412 86

View Full Product Details