Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000256.3 | NP_000247.2 | 19 - 20 | 1955 | 56 |
GenBank mRNA | AK291569.1 | - | 19 - 20 | 1955 | 56 |
AK297843.1 | - | 19 - 20 | 1955 | 56 | |
BC136543.1 | - | 19 - 20 | 1932 | 56 | |
BC136546.1 | - | 19 - 20 | 1932 | 56 | |
BC142685.1 | - | 19 - 20 | 1935 | 56 | |
BC151211.1 | - | 19 - 20 | 1938 | 56 | |
EF560722.1 | - | 19 - 20 | 1956 | 56 | |
EU747718.1 | - | 19 - 20 | 1955 | 56 | |
EU747719.1 | - | 19 - 20 | 1955 | 56 |