Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000256.3 | NP_000247.2 | 31 - 32 | 3547 | 61 |
GenBank mRNA | AK291569.1 | - | 31 - 32 | 3547 | 61 |
BC136543.1 | - | 31 - 32 | 3524 | 61 | |
BC136546.1 | - | 31 - 32 | 3524 | 61 | |
BC142685.1 | - | 31 - 32 | 3527 | 61 | |
BC151211.1 | - | 31 - 32 | 3530 | 61 | |
EF560722.1 | - | 31 - 32 | 3548 | 61 | |
EU747718.1 | - | 31 - 32 | 3547 | 61 | |
EU747719.1 | - | 31 - 32 | 3547 | 61 |