Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000256.3 | NP_000247.2 | 2 - 3 | 349 | 164 |
GenBank mRNA | AK291569.1 | - | 2 - 3 | 349 | 164 |
AK297843.1 | - | 2 - 3 | 349 | 164 | |
AU140784.1 | - | 2 - 3 | 349 | 164 | |
BC136543.1 | - | 2 - 3 | 326 | 164 | |
BC136546.1 | - | 2 - 3 | 326 | 164 | |
BC142685.1 | - | 2 - 3 | 329 | 164 | |
BC151211.1 | - | 2 - 3 | 332 | 164 | |
EU747718.1 | - | 2 - 3 | 349 | 164 | |
EU747719.1 | - | 2 - 3 | 349 | 164 |