Assay Design



Gene Transcripts

Gene Symbol
EDN3
Entrez Gene ID
1908
Gene Name
endothelin 3
Gene Aliases
ET-3, ET3, HSCR4, PPET3, WS4B
Chromosome Location
Chr.20: 59300415 - 59325992 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207032.2 NP_996915.1 4 - 5 1012 68
GenBank mRNA BC053866.1 - 4 - 5 996 68
S50864.1 - 2 - 3 275 68

View Full Product Details