Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010927.2 | NP_001010927.1 | 9 - 10 | 1084 | 105 |
NM_012454.3 | NP_036586.2 | 22 - 23 | 4332 | 105 | |
GenBank mRNA | AF120323.1 | - | 18 - 19 | 2302 | 105 |
AF120324.1 | - | 9 - 10 | 1060 | 105 | |
AF195656.1 | - | 4 - 5 | 381 | 105 | |
AK125459.1 | - | 20 - 21 | 2878 | 105 | |
AK299723.1 | - | 9 - 10 | 1081 | 105 | |
AK302860.1 | - | 18 - 19 | 2243 | 105 | |
AL122086.1 | - | 17 - 18 | 2161 | 105 | |
BC137275.1 | - | 21 - 22 | 4205 | 105 |