Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012454.3 | NP_036586.2 | 10 - 11 | 2830 | 138 |
GenBank mRNA | AF120323.1 | - | 6 - 7 | 800 | 138 |
AK123859.1 | - | 11 - 12 | 2925 | 138 | |
AK125459.1 | - | 7 - 8 | 1289 | 138 | |
AK302860.1 | - | 6 - 7 | 741 | 138 | |
AL122086.1 | - | 5 - 6 | 659 | 138 | |
BC137275.1 | - | 9 - 10 | 2703 | 138 |