Assay Design



Gene Transcripts

Gene Symbol
CNPPD1
Entrez Gene ID
27013
Gene Name
cyclin Pas1/PHO80 domain containing 1
Gene Aliases
C2orf24, CGI-57
Chromosome Location
Chr.2: 219171897 - 219178174 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321391.1 NP_001308320.1 1 - 2 88 64
NR_135628.1 1 - 2 88 64
GenBank mRNA BI912613.1 - 1 - 2 55 64

View Full Product Details