Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008405.2 | NP_001008405.1 | 1 - 2 | 111 | 135 |
NM_018844.3 | NP_061332.2 | 2 - 3 | 436 | 135 | |
XM_006716051.2 | XP_006716114.1 | 2 - 3 | 432 | 135 | |
XM_011516406.2 | XP_011514708.1 | 2 - 3 | 195 | 135 | |
XM_011516407.1 | XP_011514709.1 | 2 - 3 | 432 | 135 | |
XM_011516408.2 | XP_011514710.1 | 2 - 3 | 432 | 135 | |
XM_011516409.2 | XP_011514711.1 | 2 - 3 | 432 | 135 | |
XM_017012436.1 | XP_016867925.1 | 2 - 3 | 432 | 135 | |
XM_017012437.1 | XP_016867926.1 | 2 - 3 | 432 | 135 | |
XM_017012438.1 | XP_016867927.1 | 2 - 3 | 432 | 135 | |
GenBank mRNA | AF126020.1 | - | 2 - 3 | 143 | 135 |
AK289401.1 | - | 2 - 3 | 193 | 135 | |
BC008478.1 | - | 2 - 3 | 148 | 135 | |
BT006981.1 | - | 1 - 2 | 97 | 135 | |
BX457671.2 | - | 2 - 3 | 151 | 135 | |
BX647653.1 | - | 2 - 3 | 147 | 135 |