Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145025.1 | NP_001138497.1 | 3 - 4 | 353 | 70 |
NM_018217.2 | NP_060687.2 | 4 - 5 | 464 | 70 | |
NR_026728.1 | 3 - 4 | 680 | 70 | ||
GenBank mRNA | AK001645.1 | - | 4 - 5 | 446 | 70 |
AK023931.1 | - | 4 - 5 | 434 | 70 | |
AK096016.1 | - | 2 - 3 | 374 | 70 | |
AK293583.1 | - | 2 - 3 | 284 | 70 | |
AK298794.1 | - | 3 - 4 | 422 | 70 | |
AK300212.1 | - | 3 - 4 | 308 | 70 | |
AY358580.1 | - | 3 - 4 | 314 | 70 | |
BC001371.2 | - | 4 - 5 | 431 | 70 | |
BC016184.1 | - | 4 - 5 | 405 | 70 | |
BP297100.1 | - | 4 - 5 | 464 | 70 | |
CR457266.1 | - | 4 - 5 | 364 | 70 |